Anti-CASP7 Mouse monoclonal antibody [clone: 10-1-62]
Catalog # USBIC2087-29C6
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:caspase 7, apoptosis-related cysteine peptidase
- Antigen symbol:CASP7
- Clonality:Monoclonal
- Clone:10-1-62
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 10mM sodium phosphate, pH 7,2, 50mM sodium chloride, 0,1mM PMSF.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: Monoclonal
Clone: 22920
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human