Anti-CARD8 Rabbit polyclonal antibody
USBIC1349-05F
- Antibody type:Primary
- Antigen name:caspase recruitment domain family, member 8
- Antigen symbol:CARD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: CARD8 (caspase recruitment domain family, member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human