Anti-CA9 Mouse monoclonal antibody unconjugated [clone: 7H20]
Catalog # USBIC1105-81
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:7H20
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:59 to 414
- Storage buffer:PBS, pH 7,4, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 2-8?g/ml
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Monoclonal
Clone: 7H20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human