Anti-CA9 Rabbit monoclonal antibody unconjugated [clone: D47G3]
Catalog # USBIC1105-80K
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:D47G3
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Format:Supernatant
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, <0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:200; Immunochemistry: 1:100
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Monoclonal
Clone: D47G3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human