Anti-CA9 Rabbit polyclonal antibody unconjugated
Catalog # USBIC1105-80E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris-citrate/phophate, pH 7,5, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Rat