Anti-CA9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBIC1105-80D-PE
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifiche
Informazioni su questo articolo
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human