Anti-CA9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H20]
Catalog # USBIC1105-80
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:7H20
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Amino acid number:59 to 414
- Storage buffer:PBS, 0,5% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Monoclonal
Clone: 7H20
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human