Anti-CA6 Mouse monoclonal antibody unconjugated [clone: 9H79]
Catalog # USBIC1105-65A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase VI
- Antigen symbol:CA6
- Clonality:Monoclonal
- Clone:9H79
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:18 to 308
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 25 ?g/ml; ELISA: 0.5-1.0 ?g/ml
Type: Primary
Antigen: CA6 (carbonic anhydrase VI)
Clonality: Monoclonal
Clone: 9H79
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human