Anti-CA6 Mouse monoclonal antibody unconjugated [clone: 7H212]
Catalog # USBIC1105-65
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase VI
- Antigen symbol:CA6
- Clonality:Monoclonal
- Clone:7H212
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:18 to 308
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: CA6 (carbonic anhydrase VI)
Clonality: Monoclonal
Clone: 7H212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human