Anti-CA3 Rabbit polyclonal antibody unconjugated
Catalog # USBIC1105-43C
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:carbonic anhydrase III, muscle specific
- Antigen symbol:CA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Specifiche
Informazioni su questo articolo
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: CA3 (carbonic anhydrase III, muscle specific)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse