Anti-CA2 Rabbit polyclonal antibody unconjugated
USBIC1105-10M
- Antibody type:Primary
- Antigen name:carbonic anhydrase II
- Antigen symbol:CA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Bovine
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:50 µl
- Pk:50 µl
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine