Anti-CA1 Mouse monoclonal antibody unconjugated [clone: 9D6D7]
USBIC1105-05H
- Antibody type:Primary
- Antigen name:carbonic anhydrase I
- Antigen symbol:CA1
- Clonality:Monoclonal
- Clone:9D6D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Amino acid number:25 to 90
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:300
Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality: Monoclonal
Clone: 9D6D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human