Anti-CA1 Rat monoclonal antibody unconjugated [clone: 7H139]
Catalog # USBIC1105-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase I
- Antigen symbol:CA1
- Clonality:Monoclonal
- Clone:7H139
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:2 to 261
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality: Monoclonal
Clone: 7H139
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human