Anti-CAP1 Rabbit Polyclonal Antibody
200 µg
USBIC1075-96
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CAP1 (CAP, adenylate cyclase-associated protein 1 (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Documents
- Size:200 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Amino acid number:192 to 206
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Antigen name:CAP, adenylate cyclase-associated Protein 1 (yeast)
- Cat. No.:USBIC1075-96
- Reactivity:Mouse
- Conjugation:Unconjugated
- Antigen symbol:CAP1
- Storage buffer:0,1M Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% sodium azide, before the addition of glycerol to 30%.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



