Anti-CAM-related/Down-regulated by Oncogenes Rat Monoclonal Antibody [clone: 8H134]
USBIC1039-50
- Antibody type:Primary
- Antigen name:CAM-related/Down-regulated by Oncogenes
- Clonality:Monoclonal
- Clone:8H134
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBSwith 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: CAM-related/Down-regulated by Oncogenes
Clonality: Monoclonal
Clone: 8H134
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse