Order Entry
Italy
ContactUsLinkComponent
Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 57-24]
Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 57-24]
Catalog # USBIC1037-02M
Supplier:  US Biological
Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 57-24]
Catalog # USBIC1037-02M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Campylobacter jejuni
  • Clonality:
    Monoclonal
  • Clone:
    57-24
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG3
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Campylobacter jejuni
Clonality: Monoclonal
Clone: 57-24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: