Order Entry
Italy
ContactUsLinkComponent
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 1.A.38]
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 1.A.38]
Catalog # USBIC0016-250
Supplier:  US Biological
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 1.A.38]
Catalog # USBIC0016-250
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
  • Antigen symbol:
    ERBB2
  • Clonality:
    Monoclonal
  • Clone:
    1.A.38
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Monoclonal
Clone: 1.A.38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human