Order Entry
Italy
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide, Pro, 61-76 Mouse monoclonal antibody unconjugated [clone: 5E865]
Anti-Brain Natriuretic Peptide, Pro, 61-76 Mouse monoclonal antibody unconjugated [clone: 5E865]
Catalog # USBIB2702-04T
Supplier:  US Biological
Anti-Brain Natriuretic Peptide, Pro, 61-76 Mouse monoclonal antibody unconjugated [clone: 5E865]
Catalog # USBIB2702-04T
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide, Pro, 61-76
  • Clonality:
    Monoclonal
  • Clone:
    5E865
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    61 to 76
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide, before the addition of glycerol to 40%.
  • Storage temperature:
    Cool
  • Concentration:
    ~4,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: Brain Natriuretic Peptide, Pro, 61-76
Clonality: monoclonal
Clone: 5E865
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human