Order Entry
Italy
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide, Pro, 46-60 Mouse monoclonal antibody unconjugated [clone: 5E521]
Anti-Brain Natriuretic Peptide, Pro, 46-60 Mouse monoclonal antibody unconjugated [clone: 5E521]
Catalog # USBIB2702-04G
Supplier:  US Biological
Anti-Brain Natriuretic Peptide, Pro, 46-60 Mouse monoclonal antibody unconjugated [clone: 5E521]
Catalog # USBIB2702-04G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide, Pro, 46-60
  • Clonality:
    Monoclonal
  • Clone:
    5E521
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    46 to 60
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    500 µg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Brain Natriuretic Peptide, Pro, 46-60
Clonality: monoclonal
Clone: 5E521
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human