Anti-Borrelia burgdorferi, Outer Surface Protein B p19 Mouse monoclonal antibody unconjugated [clone: 10B313]
Catalog # USBIB2570-26P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Borrelia burgdorferi, Outer Surface Protein B p19
- Clonality:Monoclonal
- Clone:10B313
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Borrelia burgdorferi, Outer Surface Protein B p19
Clonality: monoclonal
Clone: 10B313
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: