Anti-BIRC2 Rabbit Polyclonal Antibody
Catalog # USBIB2049-90F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 2
- Antigen symbol:BIRC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Amino acid number:2 to 18
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: BIRC2 (baculoviral IAP repeat containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human