Anti-BCL2L11 Rabbit Polyclonal Antibody (Biotin)
USBIB1090-02D-BIOT
- Antibody type:Primary
- Antigen name:BCL2-like 11 (apoptosis facilitator)
- Antigen symbol:BCL2L11
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: BCL2L11 (BCL2-like 11 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human