Anti-PRKAA1 Rabbit Monoclonal Antibody [clone: 9B144]
Catalog # USBIA1475-09A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Monoclonal
- Clone:9B144
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Drosophila,Human,Rat,Mouse
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Modification:phospho-Thr172
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:2000
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: monoclonal
Clone: 9B144
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Mouse, Rat