Order Entry
Italy
ContactUsLinkComponent
 
Anti-PRKAA1 Rabbit Monoclonal Antibody Sepharose [clone: 10A2]
  USBIA1475-01M
Anti-PRKAA1 Rabbit Monoclonal Antibody Sepharose [clone: 10A2]
  USBIA1475-01M

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase, AMP-activated, alpha 1 catalytic subunit
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Monoclonal
  • Clone:
    10A2
  • Conjugation:
    Sepharose
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    400 µl
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: monoclonal
Clone: 10A2
Conjugation: Sepharose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat