Order Entry
Italy
ContactUsLinkComponent
Anti-Adenovirus Type 40-41 Mouse Monoclonal Antibody [clone: 10K4]
Anti-Adenovirus Type 40-41 Mouse Monoclonal Antibody [clone: 10K4]
Catalog # USBIA0880-45C
Supplier:  US Biological
Anti-Adenovirus Type 40-41 Mouse Monoclonal Antibody [clone: 10K4]
Catalog # USBIA0880-45C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenovirus Type 40-41
  • Clonality:
    Monoclonal
  • Clone:
    10K4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~6 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Adenovirus Type 40-41
Clonality: monoclonal
Clone: 10K4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: