Order Entry
Italy
ContactUsLinkComponent
Anti-ORM1 Rat Monoclonal Antibody [clone: 10K30]
Anti-ORM1 Rat Monoclonal Antibody [clone: 10K30]
Catalog # USBIA0550-01M
Supplier:  US Biological
Anti-ORM1 Rat Monoclonal Antibody [clone: 10K30]
Catalog # USBIA0550-01M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    orosomucoid 1
  • Antigen symbol:
    ORM1
  • Clonality:
    Monoclonal
  • Clone:
    10K30
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: ORM1 (orosomucoid 1)
Clonality: monoclonal
Clone: 10K30
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse