Anti-ANAPC1 Rabbit Polyclonal Antibody
Catalog # USBI168019
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Anaphase Promoting Complex Subunit 1
- Antigen symbol:ANAPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:APC1
- Storage buffer:0,02M potassium phosphate, 0,15M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:5000; Immunoprecipitation: 1:100
Type: Primary
Antigen: ANAPC1 (anaphase promoting complex subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human