Order Entry
Italy
ContactUsLinkComponent
 
Anti-SLC18A3 Mouse Monoclonal Antibody [clone: S6-38]
  USBI167615
Anti-SLC18A3 Mouse Monoclonal Antibody [clone: S6-38]
  USBI167615

 

  • Antibody type:
    Primary
  • Antigen name:
    Solute carrier family 18 (vesicular acetylcholine transporter), member 3
  • Antigen symbol:
    SLC18A3
  • Clonality:
    Monoclonal
  • Clone:
    S6-38
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBSpH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SLC18A3 (solute carrier family 18 (vesicular acetylcholine transporter), member 3)
Clonality: monoclonal
Clone: S6-38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat