Order Entry
Italy
ContactUsLinkComponent
Anti-Copper Transporting ATPase2 Mouse Monoclonal Antibody [clone: S62-29]
Anti-Copper Transporting ATPase2 Mouse Monoclonal Antibody [clone: S62-29]
Catalog # USBI167582
Supplier:  US Biological
Anti-Copper Transporting ATPase2 Mouse Monoclonal Antibody [clone: S62-29]
Catalog # USBI167582
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Copper Transporting ATPase2
  • Clonality:
    Monoclonal
  • Clone:
    S62-29
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBSpH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Copper Transporting ATPase2
Clonality: monoclonal
Clone: S62-29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat