Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-17]
Catalog # USBI167576
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ankyrin 2, neuronal
- Antigen symbol:ANK2
- Clonality:Monoclonal
- Clone:S105-17
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: ANK2 (ankyrin 2, neuronal)
Clonality: monoclonal
Clone: S105-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat