Anti-ADRA2C Mouse Monoclonal Antibody [clone: S330A-51]
Catalog # USBI167571
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Adrenoceptor alpha 2C
- Antigen symbol:ADRA2C
- Clonality:Monoclonal
- Clone:S330A-51
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: ADRA2C (adrenoceptor alpha 2C)
Clonality: monoclonal
Clone: S330A-51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse