Order Entry
Italy
ContactUsLinkComponent
Anti-ADAM22 Mouse Monoclonal Antibody [clone: S46-30]
Anti-ADAM22 Mouse Monoclonal Antibody [clone: S46-30]
Catalog # USBI167569
Supplier:  US Biological
Anti-ADAM22 Mouse Monoclonal Antibody [clone: S46-30]
Catalog # USBI167569
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAM metallopeptidase domain 22
  • Antigen symbol:
    ADAM22
  • Clonality:
    Monoclonal
  • Clone:
    S46-30
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBSpH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: ADAM22 (ADAM metallopeptidase domain 22)
Clonality: monoclonal
Clone: S46-30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat