Order Entry
Italy
ContactUsLinkComponent
 
Anti-TP53 Mouse Monoclonal Antibody [clone: IMD-53]
  USBI145026
Anti-TP53 Mouse Monoclonal Antibody [clone: IMD-53]
  USBI145026

 

  • Antibody type:
    Primary
  • Antigen name:
    Tumor protein p53
  • Antigen symbol:
    P53
  • Clonality:
    Monoclonal
  • Clone:
    IMD-53
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: IMD-53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human