Order Entry
Italy
ContactUsLinkComponent
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Catalog # USBI145022
Supplier:  US Biological
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Catalog # USBI145022
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase inhibitor 1B
  • Antigen symbol:
    CDKN1B
  • Clonality:
    Monoclonal
  • Clone:
    IMD-27
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: monoclonal
Clone: IMD-27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat