Order Entry
Italy
ContactUsLinkComponent
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Catalog # USBI144417
Supplier:  US Biological
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Catalog # USBI144417
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cyclin A2
  • Antigen symbol:
    CCNA2
  • Clonality:
    Monoclonal
  • Clone:
    CY-28
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Bovine,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: monoclonal
Clone: CY-28
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mouse