Anti-MADCAM1 Rat Monoclonal Antibody [clone: 13H302]
USBI138109
- Antibody type:Primary
- Antigen name:Mucosal Addressin Cell Adhesion Molecule 1
- Antigen symbol:MADCAM1
- Clonality:Monoclonal
- Clone:13H302
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:100mM BBS, pH 8,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: MADCAM1 (mucosal vascular addressin cell adhesion molecule 1)
Clonality: monoclonal
Clone: 13H302
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse