Order Entry
Italy
ContactUsLinkComponent
Anti-Serotonin Transporter Mouse Monoclonal Antibody [clone: 17-7A4]
Anti-Serotonin Transporter Mouse Monoclonal Antibody [clone: 17-7A4]
Catalog # USBI136958
Supplier:  US Biological
Anti-Serotonin Transporter Mouse Monoclonal Antibody [clone: 17-7A4]
Catalog # USBI136958
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Serotonin Transporter
  • Clonality:
    Monoclonal
  • Clone:
    17-7A4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Serotonin Transporter
Clonality: monoclonal
Clone: 17-7A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat