Anti-ZNF79 Mouse Monoclonal Antibody [clone: 3D12]
Catalog # USBI135804
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 79
- Antigen symbol:ZNF79
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:WRCLVSTPRDRFKEGIPGKSRSLVLLGLPVSQPGMNSQLEQREGAWMLEGEDLRSPSPGWKIISGSPPEQALSEASFQDPCVEMPPGDSDHGTSDLEKSF*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZNF79 (zinc finger protein 79)
Clonality: monoclonal
Clone: 3D12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human