Anti-ZNF69 Mouse Monoclonal Antibody [clone: 1E3]
Catalog # USBI135788
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 69
- Antigen symbol:ZNF69
- Clonality:Monoclonal
- Clone:1E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:1 to 82
- Storage buffer:PBS, pH 7,2.
- Sequence:MPCCSHRRCREDPGTSESQEMEEWALLDISQRKLYKEVMLETFRNLTSVGKSWKDQNIEYEYQNPRRNFRSLIEKKVNEIK*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZNF69 (zinc finger protein 69)
Clonality: monoclonal
Clone: 1000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human