Anti-ZFHX3 Mouse Monoclonal Antibody [clone: 3B1]
Catalog # USBI135543
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger homeobox 3
- Antigen symbol:ZFHX3
- Clonality:Monoclonal
- Clone:3B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:2811 to 2910
- Storage buffer:PBS, pH 7,2.
- Sequence:EDFESPSMSSVNLNFDQTKLDNDDCSSVNTAITDTTTGDEGNADNDSATGIATETKSSSAPNEGLTKAAMMAMSEYEDRLSSGLVSPAPSFYSKEYDNEG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: ZFHX3 (zinc finger homeobox 3)
Clonality: monoclonal
Clone: 3B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human