Order Entry
Italy
ContactUsLinkComponent
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 6A10]
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 6A10]
Catalog # USBI135488
Supplier:  US Biological
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 6A10]
Catalog # USBI135488
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 gamma
  • Clonality:
    Monoclonal
  • Clone:
    6A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    70 to 170
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    TSADGNEKKIEMVRAYREKIEKELEAVCQDVLSLLDNYLIKNCSETQYESKVFYLKMKGDYYRYLAEVATGEKRATVVESSEKAYSEAHEISKEHMQPTH*
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: 14-3-3 gamma (14-3-3 gamma)
Clonality: monoclonal
Clone: 6A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat