Order Entry
Italy
ContactUsLinkComponent
Anti-RING Finger Protein 168 Mouse Monoclonal Antibody [clone: 3E1]
Anti-RING Finger Protein 168 Mouse Monoclonal Antibody [clone: 3E1]
  USBI132669
Anti-RING Finger Protein 168 Mouse Monoclonal Antibody [clone: 3E1]
  USBI132669

 

  • Antibody type:
    Primary
  • Antigen name:
    Ring Finger Protein 168
  • Antigen symbol:
    RNF168
  • Clonality:
    Monoclonal
  • Clone:
    3E1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    462 to 572
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MVPNRQKGSPDEYHLRATSSPPDKVLNGQRKNPKDGNFKRQTHTKHPTPERGSRDKNRQVSLKMQLKQSVNRRKMPNSTRDHCKVSKSAHSLQPSISQKSVFQMFQRCTK
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: RING Finger Protein 168
Clonality: monoclonal
Clone: 30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human