Anti-Retinol Dehydrogenase 12 Mouse Monoclonal Antibody [clone: 1E11]
Catalog # USBI132442
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Retinol Dehydrogenase 12
- Antigen symbol:RDH12
- Clonality:Monoclonal
- Clone:1E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:217 to 317
- Storage buffer:PBS, pH 7,2.
- Sequence:RLQGTGVTTYAVHPGVVRSELVRHSSLLCLLWRLFSPFVKTAREGAQTSLHCALAEGLEPLSGKYFSDCKRTWVSPRARNNKTAERLWNVSCELLGIRWE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Retinol Dehydrogenase 12
Clonality: monoclonal
Clone: 100000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human