Anti-RASGRP3 Mouse Monoclonal Antibody [clone: 1H4]
USBI132346
- Antibody type:Primary
- Antigen name:RAS guanyl releasing protein 3 (calcium and DAG-regulated)
- Antigen symbol:RASGRP3
- Clonality:Monoclonal
- Clone:1H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:1 to 99
- Storage buffer:PBS, pH 7,2.
- Sequence:MGSSGLGKAATLDELLCTCIEMFDDNGELDNSYLPRIVLLMHRWYLSSTELAEKLLCMYRNATGESCNEFRLKICYFMRYWILKFPAEFNLDLGLIRMT*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: RASGRP3 (RAS guanyl releasing protein 3 (calcium and DAG-regulated))
Clonality: monoclonal
Clone: 1H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human