Order Entry
Italy
ContactUsLinkComponent
 
Anti-PYCR1 Rabbit Polyclonal Antibody
  USBI132127
Anti-PYCR1 Rabbit Polyclonal Antibody
  USBI132127

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyrroline-5-carboxylate reductase 1
  • Antigen symbol:
    PYCR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    Serum
  • Form:
    liquid
  • Amino acid number:
    1 to 319
  • Sequence:
    MSVGFIGAGQLAFALAKGFTAAGVLAAHKIMASSPDMDLATVSALRKMGVKLTPHNKETVQHSDVLFLAVKPHIIPFILDEIGADIEDRHIVVSCAAGVTISSIEKKLSAFRPAPRVIRCMTNTPVVVREGATVYATGTHAQVEDGRLMEQLLSSVGFCTEVEEDLIDAVTGLSGSGPAYAFTALDALADGGVKMGLPRRLAVRLGAQALLGAAKMLLHSEQHPGQLKDNVSSPGGATIHALHVLESGGFRSLLINAVEASCIRTRELQSMADQEQVSPAAIKKTILDKVKLDSPAGTALSPSGHTKLLPRSLAPAGKD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: PYCR1 (pyrroline-5-carboxylate reductase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human