Anti-PRKCI Mouse Monoclonal Antibody [clone: 1G11]
Catalog # USBI131805
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase C Iota
- Antigen symbol:PRKCI
- Clonality:Monoclonal
- Clone:1G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: PRKCI (protein kinase C, iota)
Clonality: monoclonal
Clone: 1G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human