Anti-PKD2 Mouse Monoclonal Antibody [clone: 4C8]
USBI131388
- Antibody type:Primary
- Antigen name:Polycystic Kidney Disease 2
- Antigen symbol:PKD2
- Clonality:Monoclonal
- Clone:4C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:261 to 361
- Storage buffer:PBS, pH 7,2.
- Sequence:PVSKTEKTNFKTLSSMEDFWKFTEGSLLDGLYWKMQPSNQTEADNRSFIFYENLLLGVPRIRQLRVRNGSCSIPQDLRDEIKECYDVYSVSSEDRAPFGP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: PKD2 (polycystic kidney disease 2 (autosomal dominant))
Clonality: monoclonal
Clone: 4C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human