Anti-NR4A2 Mouse Monoclonal Antibody [clone: 4A6]
Catalog # USBI130558
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:nuclear receptor subfamily 4, group A, member 2
- Antigen symbol:NR4A2
- Clonality:Monoclonal
- Clone:4A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:GFQVQHSPMWDDPGSLHNFHQNYVATTHMIEQRKTPVSRLSLFSFKQSPPGTPVSSCQMRFDGPLHVPMNPEPAGSHHVVDGQTFAVPNPIRKPASMGFP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: NR4A2 (nuclear receptor subfamily 4, group A, member 2)
Clonality: monoclonal
Clone: 4A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human