Anti-Nociceptin Receptor Mouse Monoclonal Antibody [clone: 2A11]
Catalog # USBI130449
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nociceptin Receptor
- Antigen symbol:NOP
- Clonality:Monoclonal
- Clone:2A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:DILLGFWPFGNALCKTVIAIDYYNMFTSTFTLTAMSVDRYVAICHPIRALDVRTSSKAQAVNVAIWALASVVGVPVAIMGSAQVEDEEIECLVEIPTPQDYWG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Nociceptin Receptor
Clonality: monoclonal
Clone: 2A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human