Anti-HSD17B8 Mouse Monoclonal Antibody [clone: 4F1]
USBI128101
- Antibody type:Primary
- Antigen name:hydroxysteroid (17-beta) dehydrogenase 8
- Antigen symbol:HSD17B8
- Clonality:Monoclonal
- Clone:4F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:28 to 128
- Storage buffer:PBS, pH 7,2.
- Sequence:SVRLAGEGATVAACDLDRAAAQETVRLLGGPGSKEGPPRGNHAAFQADVSEARAARCLLEQVQACFSRPPSVVVSCAGITQDEFLLHMSEDDWDKVIAVN*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HSD17B8 (hydroxysteroid (17-beta) dehydrogenase 8)
Clonality: monoclonal
Clone: 4F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human